Learn More
IKA™ R 1376 Paddle Stirrer
Overhead, paddle-shaped stirring element for gentle mixing at low to medium speeds.
Marca: IKA™ 0000757800
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- Tangential flow
- Minimum turbulence
- Good heat exchange
- Gentle treatment of product
- Use at low to medium speeds
Specifica
Model R 1376 | |
0.4 in. | |
10 mm | |
21.7 in. | |
800 rpm |
5.9 in. | |
150 mm | |
Gentle mixing at low to medium speeds | |
550 mm | |
Paddle Stirrer |