Learn More
Abnova™ PALB2/CEN16q FISH Probe
Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique.
Marca: Abnova™ FG0182.100uL
Questo articolo non è restituibile.
Consulta la politica di reso
Specifica
Store at 4°C in the dark. | |
Tube | |
79728 | |
100 μL |
DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe | |
FISH-Ce | |
DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe | |
Liquid |