missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbuterol acetate, Honeywell Fluka™
Click to view available options
Quantità:
10 mg
Descrizione
Ensure you have the information you need to quickly start your analysis, and pass relevant audits:
- High quality inorganic and organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results
- Free expert technical support
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifica
Forma fisica | Crystalline |
CAS | 1613439-57-8 |
Grado | Analytical Standard |
Imballaggio | Glass Bottle |
Quantità | 10 mg |
Da utilizzare con (applicazione) | Beta-agonists and Antagonists |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'