missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals PARP Inhibitor VIII, PJ34
Identificatori chimici
CAS | 344458-15-7 |
---|---|
Formula molecolare | C17H17N3O2·HCl, C17H18ClN3O2 |
Sinonimo | 2-Dimethylamino-N-(6-oxo-5,6-dihydrophenanthridin-2-yl)acetamide hydrochloride |
Descrizione
A cell-permeable, water-soluble phenanthridinone-derivative that acts as a potent inhibitor of poly(ADP-ribose) polymerase
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- A cell-permeable, water-soluble phenanthridinone-derivative that acts as a potent inhibitor of poly(ADP-ribose) polymerase
Identificatori chimici
344458-15-7 | |
C17H17N3O2·HCl, C17H18ClN3O2 | |
2-Dimethylamino-N-(6-oxo-5,6-dihydrophenanthridin-2-yl)acetamide hydrochloride |
Specifica
White | |
A cell-permeable, water-soluble phenanthridinone-derivative that acts as a potent inhibitor of poly(ADP-ribose) polymerase | |
C17H17N3O2·HCl, C17H18ClN3O2 | |
2-Dimethylamino-N-(6-oxo-5,6-dihydrophenanthridin-2-yl)acetamide hydrochloride | |
Signal Transduction Reagent | |
RURAZZMDMNRXMI-UHFFFAOYSA-N | |
2-(dimethylamino)-N-(6-oxo-5,6-dihydrophenanthridin-2-yl)acetamide hydrochloride |
-30°C to -10°C | |
344458-15-7 | |
331.80g/mol | |
Light Sensitive and Hygroscopic | |
Solid | |
Cl.CN(C)CC(=O)NC1=CC=C2NC(=O)C3=CC=CC=C3C2=C1 | |
331.80 |
Sicurezza e movimentazione
Recommended Storage : Store at -20°C
RUO – Research Use Only