Learn More
DiscoverX PathHunter™ C3AR1 Activated GPCR Internalization U2OS Cell Line
The PathHunter™ C3AR1 Activated GPCR Internalization U2OS Cell Line measures C3AR1 (GPCR) internalization via early-endosome localization. This cell line ships as two vials of cryopreserved cells.
Marca: DiscoverX 93-0771C3
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
The PathHunter™ C3AR1 Activated GPCR internalization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) affecting GPCR activation and internalization. This cell line enables assessment of ligand (e.g. C3a) based internalization of C3AR1 via detection of localization to the early endosome. This product uses EFC technology. The included cell line overexpresses EA-tagged β-arrestin 2 as well as a PK-tagged early endosome protein and untagged C3AR1. Upon ligand-induced receptor activation, β-arrestin 2 is recruited to the activated GPCR and the resulting C3AR1-arrestin complex is internalized to the early endosome, leading to production of EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells.Specifica
Receptor Internalization | |
Cell Lines | |
2 Vials | |
NM_004054.2 |
U2OS | |
C3a Receptor Agonist | |
C3AR1 |
Il programma Encompass di Fisher Scientific offre articoli che non fanno parte del nostro portafoglio di distribuzione. Questi prodotti in genere non hanno immagini o descrizioni dettagliate. Tuttavia, ci impegniamo a migliorare la vostra esperienza di acquisto. Si prega di utilizzare il modulo sottostante per fornire un feedback relativo al contenuto di questo prodotto.