Learn More
DiscoverX PathHunter™ CHO-K1 ADCYAP1R1 (PAC1) β-Arrestin Cell Line
The PathHunter™ CHO-K1 ADCYAP1R1 (PAC1) β-Arrestin Cell Line measures ADCYAP1R1 (GPCR) activity via recruitment of β-Arrestin2. This cell line ships as two vials of cryopreserved cells.
Marca: DiscoverX 93-0249C2
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
The PathHunter™ ADCYAP1R1 (PAC1) β-Arrestin cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. MECA) based activation of ADCYAP1R1 GPCR activity via detection of β-arrestin recruitment. This product uses EFC technology (www.discoverx.com/technologies-platforms/enzyme-fragment-complementation-technology). The included cell line overexpresses PK-tagged ADCYAP1R1 and EA-tagged β-Arrestin 2. Activation of ADCYAP1R1 stimulates the recruitment of β-Arrestin 2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells.Specifica
CHO-K1 | |
PACAP (1-27) | |
ADCYAP1R1 (PAC1) |
Cell Lines | |
2 Vials | |
AY366498 |