Learn More
DiscoverX PathHunter™ CHO-K1 AR Protein Interaction Cell Line
The PathHunter™ CHO-K1 AR Protein Interaction Cell Line measures activated AR nuclear hormone receptor interaction with SRCPs. This cell line ships as two vials of cryopreserved cells.
Marca: DiscoverX 93-0454C2
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
The PathHunter™ AR Protein Interaction cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) affecting NHR activation and recruitment of cofactors. This cell line enables assessment of ligand (e.g. testosterone, norgestrel) based NHR activation via detection of SRCP (steroid receptor co-activator peptide) recruitment. This product uses EFC technology. The included cell line overexpresses PK-tagged AR NHR and EA-tagged steroid receptor co-activator peptide (SRCP). Upon activation of AR, induced interaction with SRCP leads to production of EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells.Specifica
Protein Interaction | |
Cell Lines | |
2 Vials | |
NM_000044 |
CHO-K1 | |
D(-)-Norgestrel | |
AR |