Learn More
DiscoverX PathHunter™ CHO-K1 CRTH2 β-Arrestin Cell Line
The PathHunter™ CHO-K1 CRTH2 β-Arrestin Cell Line measures CRTH2 (GPCR) activity via recruitment of β-Arrestin2. This cell line ships as two vials of cryopreserved cells.
Marca: DiscoverX 93-0291C2
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
The PathHunter™ CRTH2 β-Arrestin cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. prostaglandin D2) based activation of CRTH2 GPCR activity via detection of β-arrestin recruitment. This product uses EFC technology (www.discoverx.com/technologies-platforms/enzyme-fragment-complementation-technology). The included cell line overexpresses PK-tagged CRTH2 and EA-tagged β-Arrestin 2. Activation of CRTH2 stimulates the recruitment of β-Arrestin 2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells.Specifica
β-Arrestin recruitment | |
Cell Lines | |
2 Vials | |
AY507142 |
CHO-K1 | |
Prostaglandin D2 | |
CRTH2 |