Learn More
DiscoverX PathHunter™ CHO-K1 GPR132 β-Arrestin Orphan GPCR Cell Line
The PathHunter™ CHO-K1 GPR132 β-Arrestin Orphan GPCR Cell Line measures GPR132 (GPCR) activity via recruitment of β-Arrestin2. This cell line ships as two vials of cryopreserved cells.
Marca: DiscoverX 93-0411C2A
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
The PathHunter™ GPR132 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand based activation of GPR132 GPCR activity via detection of β-arrestin recruitment. This product uses EFC technology (www.discoverx.com/technologies-platforms/enzyme-fragment-complementation-technology). The included cell line overexpresses PK-tagged GPR132 and EA-tagged β-Arrestin 2. Activation of GPR132 stimulates the recruitment of β-Arrestin 2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. Orphan receptor with unknown ligand.Specifica
β-Arrestin recruitment | |
Cell Lines | |
GPR132 |
CHO-K1 | |
2 Vials | |
NM_013345 |