Learn More
DiscoverX PathHunter™ CHO-K1 LGR5 (GPR49) β-Arrestin Orphan GPCR Cell Line
The PathHunter™ CHO-K1 LGR5 (GPR49) β-Arrestin Orphan GPCR Cell Line measures LGR5 (GPCR) activity via recruitment of β-Arrestin2. This cell line ships as two vials of cryopreserved cells.
Marca: DiscoverX 93-0526C2A
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
The PathHunter™ LGR5 (GPR49) cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. R-spondins) based activation of LGR5 GPCR activity via detection of β-arrestin recruitment. This product uses EFC technology (www.discoverx.com/technologies-platforms/enzyme-fragment-complementation-technology). The included cell line overexpresses PK-tagged LGR5 and EA-tagged β-Arrestin 2. Activation of LGR5 stimulates the recruitment of β-Arrestin 2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells.Specifica
β-Arrestin recruitment | |
2 vials | |
2 Vials | |
NM_003667 |
CHO-K1 | |
Cell Lines | |
LGR5 (GPR49) |