Learn More
DiscoverX PathHunter™ CHO-K1 mCCR5 β-Arrestin Cell Line
The PathHunter™ CHO-K1 mCCR5 β-Arrestin Cell Line measures mCCR5 (GPCR) activity via recruitment of β-Arrestin2. This cell line ships as two vials of cryopreserved cells. Mouse protein.
Marca: DiscoverX 93-0470C2
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
The PathHunter™ mCCR5 β-Arrestin cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. CCL3/MIP-1α) based activation of mCCR5 GPCR activity via detection of β-arrestin recruitment. This product uses EFC technology (www.discoverx.com/technologies-platforms/enzyme-fragment-complementation-technology). The included cell line overexpresses PK-tagged mCCR5 and EA-tagged β-Arrestin 2. Activation of mCCR5 stimulates the recruitment of β-Arrestin 2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. Please note that this product expresses the Mouse version of this protein.Specifica
β-Arrestin recruitment | |
Cell Lines | |
2 Vials | |
NM_009917 |
CHO-K1 | |
Recombinant Murine MIP-1α (CCL3) | |
mCCR5 |