Learn More
DiscoverX PathHunter™ CHO-K1 MRGPRE β-Arrestin Orphan GPCR Cell Line
The PathHunter™ CHO-K1 MRGPRE β-Arrestin Orphan GPCR Cell Line measures MRGPRE (GPCR) activity via recruitment of β-Arrestin2. This cell line ships as two vials of cryopreserved cells.
Marca: DiscoverX 93-0438C2A
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
The PathHunter™ MRGPRE cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand based activation of MRGPRE GPCR activity via detection of β-arrestin recruitment. This product uses EFC technology (www.discoverx.com/technologies-platforms/enzyme-fragment-complementation-technology). The included cell line overexpresses PK-tagged MRGPRE and EA-tagged β-Arrestin 2. Activation of MRGPRE stimulates the recruitment of β-Arrestin 2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. Orphan receptor with unknown ligand.Specifica
β-Arrestin recruitment | |
Cell Lines | |
MRGPRE |
CHO-K1 | |
2 Vials | |
NM_001039165.2 |