Learn More
DiscoverX PathHunter™ CHO-K1 RARβ Protein Interaction Cell Line
The PathHunter™ CHO-K1 RARβ Protein Interaction Cell Line measures activated RARβ nuclear hormone receptor interaction with SRCPs. This cell line ships as two vials of cryopreserved cells.
Marca: DiscoverX 93-0326C2
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
The PathHunter™ RARβ Protein Interaction cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) affecting NHR activation and recruitment of cofactors. This cell line enables assessment of ligand (e.g. retinoic acid) based NHR activation via detection of SRCP (steroid receptor co-activator peptide) recruitment. This product uses EFC technology. The included cell line overexpresses PK-tagged RARβ NHR and EA-tagged steroid receptor co-activator peptide (SRCP). Upon activation of RARβ, induced interaction with SRCP leads to production of EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells.Specifica
Protein Interaction | |
Cell Lines | |
2 Vials | |
NM_000965 |
CHO-K1 | |
9-cis-Retinoic acid | |
RARβ |