Learn More
DiscoverX PathHunter™ CHO-K1 rCHRM4 β-Arrestin Cell Line
The PathHunter™ CHO-K1 rCHRM4 β-Arrestin Cell Line measures rCHRM4 (GPCR) activity via recruitment of β-Arrestin2. This cell line ships as two vials of cryopreserved cells. Rat protein.
Marca: DiscoverX 93-0870C2
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
The PathHunter™ rCHRM4 β-Arrestin cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. acetylcholine) based activation of rCHRM4 GPCR activity via detection of β-arrestin recruitment. This product uses EFC technology (www.discoverx.com/technologies-platforms/enzyme-fragment-complementation-technology). The included cell line overexpresses PK-tagged rCHRM4 and EA-tagged β-Arrestin 2. Activation of rCHRM4 stimulates the recruitment of β-Arrestin 2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. Please note that this product expresses the rat version of this protein.Specifica
β-Arrestin Recruitment | |
Cell Lines | |
2 Vials | |
NM_031547 |
CHO-K1 | |
Acetylcholine | |
rCHRM4 |