Learn More
DiscoverX PathHunter™ U2OS BMPR1A/BMPR2 Dimerization Cell Line
The PathHunter™ U2OS BMPR1A/BMPR2 Dimerization Cell Line measures ligand-induced dimerization of BMPR1A/BMPR2 . This cell line ships as two vials of cryopreserved cells.
Marca: DiscoverX 93-1006C3
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
The PathHunter™ BMPR1A/BMPR2 Dimerization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This cell line enables assessment of ligand (e.g. BMP-4) induced dimerization of BMPR1A/BMPR2 receptors. This product uses EFC technology. The cell line included overexpresses BMPR2 target protein fused to PK alongside BMPR1A target protein fused to EA. Upon ligand-induced dimerization of BMPR1A/BMPR2 , detectable EFC signal is produced. This stable modified cell line is provided as two vials of cryopreserved cells.Specifica
Dimerization | |
Cell Lines | |
2 Vials | |
NM_004329.2 |
U2OS | |
BMP-4 | |
BMPR1A/BMPR2 |