Learn More
DiscoverX PathHunter™ U2OS PRLR-JAK2 Functional Assay
The PathHunter™ U2OS PRLR-JAK2 Functional Assay measures PRLR cytokine receptor activity via ligand-induced SH2 domain protein recruitment. This cell line ships as two vials of cryopreserved cells.
Marca: DiscoverX 93-0687C3
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
The PathHunter™ PRLR-JAK2 Functional assay cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting CTKs. This cell line enables assessment of PRLR activation via detection of JAK2 mediated phosphorylation-induced SH2 domain protein recruitment. This product uses EFC technology. The included cell line overexpresses PK-tagged PRLR cytokine receptor, an untagged CTK JAK2 and EA-tagged SH2 domain protein. Upon activation of the cytokine receptor, CTK-based receptor phosphorylation leads to SH2-EA recruitment and production of EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells.Specifica
CTK Activation | |
Cell Lines | |
2 Vials | |
NM_000949 |
U2OS | |
Recombinant Human Prolactin | |
PRLR-JAK2 |