missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Pazopanib

Catalog Number p-7214410
Click to view available options
Quantità:
1 g
5 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
444731-52-6
C21H23N7O2S
437.52
CUIHSIWYWATEQL-UHFFFAOYSA-N
5-({4-[(2,3-dimethyl-2H-indazol-6-yl)(methyl)amino]pyrimidin-2-yl}amino)-2-methylbenzene-1-sulfonamide
CN(C1=CC2=NN(C)C(C)=C2C=C1)C1=NC(NC2=CC(=C(C)C=C2)S(N)(=O)=O)=NC=C1
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
444731-52-6
C21H23N7O2S
437.52
CUIHSIWYWATEQL-UHFFFAOYSA-N
5-({4-[(2,3-dimethyl-2H-indazol-6-yl)(methyl)amino]pyrimidin-2-yl}amino)-2-methylbenzene-1-sulfonamide
CN(C1=CC2=NN(C)C(C)=C2C=C1)C1=NC(NC2=CC(=C(C)C=C2)S(N)(=O)=O)=NC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Materiale o nome chimico Pazopanib
Punto di fusione 285.0°C to 289.0°C
Colore Clear
Quantità 1 g
Peso formulazione 437.52

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'