Learn More
PDCD1 / PD1 / CD279 Rabbit, Clone: PDCD1/1410R, enQuireBio™
Rabbit Monoclonal Antibody
Marca: EnquireBio 5133-RBM4-P0
Questo articolo non è restituibile.
Consulta la politica di reso
Specifica
| PDCD1 / PD1 / CD279 | |
| Cell Surface and Cytoplasmic | |
| PDCD1/1410R | |
| Unconjugated | |
| NP_016859782 | |
| Rabbit | |
| 55kDa | |
| Protein A/G Purified | |
| RUO | |
| 5133 | |
| IgG |
| Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with or without 0.05% BSA and 0.05% azide. | |
| PDCD1 | |
| Human recombinant PDCD1 protein fragment | |
| TY cells or Tonsil. | |
| 20 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |