missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ PELTOR™ FLX2 Cable for CH-3 Listen Only Headset
Click to view available options
Modello:
FLX2-203
FLX2-205
Da utilizzare con (apparecchiatura):
2.5 mm mono plug
3.5 mm mono plug
Descrizione
- For use with CH-3 Listen-Only Headsets
- Simple and reliable connection to a wide variety of handheld devices.
- Coiled polyurethane cable
- Aramid fiber reinforced
- Robust and highly flexible
Compliance
3M™ ear muffs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Specifica
Colore | Black |
Da utilizzare con (apparecchiatura) | 3.5 mm mono plug |
Da utilizzare con (applicazione) | Enables connection to listen-only CH-3 Headsets with Hytera, Icom, Kenwood or Motorola radios |
Modello | FLX2-205 |
Pairs per Package | 1 Ea. |
Product Type | FLX2 Cable |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'