Learn More
3M™ Peltor™ Short Peak Faceshield
Click to view available options
Color: Clear
Descrizione
- Clear polycarbonate visor
- Thickness: 1.2 mm
- Temperature range: -40 to 100°C
- Face shields are designed to fit 3M™ Hard Hats
- The visor meets the Grade F requirement for low energy impact protective shields
- Marking Class 3-1.2 8 9 1F