Learn More
Testo™ Type T Super-Quick Needle Probe
For measurements without visible penetration hole.
Marca: Testo™ 0628 0027
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- Ideal for hamburgers, steaks, etc.
- Waterproof
- Reaction time of 2 seconds
- Measuring range of −50° to +250°C
Specifica
1.4 mm | |
Reaction time of 2 seconds | |
-60°C | |
±0.2°C from −20° to +70°C |
150 mm | |
250°C | |
-50°C to +250°C |