Learn More
SI Analytics™ Micro-elettrodo pH combinato con cavo fisso da 1 m
Codice prodotto. 10007900 Sfoglia Tutto SI Analytics Prodotti
Click to view available options
Tipi di connettore:
DIN
DIN, 4 mm Banana Plug
Plug Head
spina BNC
Tipo:
Combination pH Micro Electrode
pH Combination Electrode
Membrana:
Cylinder
Spear
Sphere
Diametro (sistema metrico):
12/3 mm
3 mm
5 mm
Dimensione della confezione:
Pezzo
Codice prodotto. 10007900
Marca: SI Analytics™ 285105151
Ensure high measurement accuracy, stability, and long service life while maintaining high adaptability to your measurement tasks. SI Analytics™ pH Combination Micro-Electrode is robust, durable, and easy to clean.
  • Proven high-end electrodes for demanding measurement tasks
  • Double junction Silamid™ reference system for fast and stable acquiring of measured values and for longer electrode life
  • Utmost versatility of pH electrodes is achieved by a large selection of junctions, membrane glass types and shapes, shaft lengths and diameters, ground joints, plug connections and integrated temperature sensors
  • Each pH combination electrode comes with individual serial number and quality certificate
TRUSTED_SUSTAINABILITY
Specifica
Tipi di connettore DIN
Giunzione Platinum Diaphragm
Materiale A-Glass
Intervallo di pH 7.0 ±0.3
Tipo Combination pH Micro Electrode
Riferimento interno Ag/AgCl
Membrana Cylinder
Intervallo di temperatura (sistema metrico) -5 to +100°C
Lunghezza (sistema metrico) 96 mm
Lunghezza (sistema metrico) cavo 1 m
Diametro (sistema metrico) 3 mm