Learn More
Picropodophyllin, MedChemExpress
MedChemExpress Picropodophyllin (AXL1717) is a selective insulin-like growth factor-1 receptor (IGF-1R) inhibitor with an IC50 of 1 nM.
Marca: Medchem Express HY-15494
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Picropodophyllin (AXL1717) is a selective insulin-like growth factor-1 receptor (IGF-1R) inhibitor with an IC50 of 1 nM.Specifica
Picropodophyllin | |
White | |
C22H22O8 | |
AXL1717 Picropodophyllin PPP | |
O=C1OC[C@]2([H])[C@@H](O)C3=C(C=C4OCOC4=C3)[C@@H](C5=CC(OC)=C(OC)C(OC)=C5)[C@@]21[H] | |
414.41 | |
Research | |
Cancer-Kinase/protease |
477-47-4 | |
Research | |
100 mg | |
DMSO : 50 mg/mL (120.65 mM; Need ultrasonic) | |
414.41 | |
98.87% | |
Solid |