Learn More
Thermo Scientific™ Pierce™ Premium Grade Sulfo-NHS-LC-Biotin
Optimize applications where product integrity and risk minimization are paramount with our highest quality formulation of this popular amine-reactive biotinylation reagent.
Marca: Thermo Scientific™ PG82076
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
Optimize applications where product integrity and risk minimization are paramount with the specially characterized Thermo Scientific™ Pierce™ Premium Grade Sulfo-NHS-LC-Biotin, our highest quality formulation of this popular amine-reactive biotinylation reagent.
Thermo Scientific Pierce Premium Grade Sulfo-NHS-LC-Biotin is our highest quality formulation of this popular amine-reactive biotinylation reagent, specially characterized for applications where product integrity and risk minimization are paramount.
Compared to the standard grade product, Premium Grade Sulfo-NHS-LC-Biotin provides more clearly defined quality and product support by including (a) increased analytical testing and product characterization, (b) greater batch-specific information and quality assurance review, (c) extensive lot sample retention and (d) change control notification. Sulfo-NHS-LC-Biotin is an intermediate-length, water-soluble biotinylation reagent for labeling antibodies, proteins and other molecules that have primary amines. Specific labeling of cell surface proteins is another common application for these uniquely water-soluble and membrane impermeable reagents.
Highlights:- Sulfo-NHS-LC-Biotin – popular amine-reactive biotinylation reagent for antibody labeling
- High quality – identity and purity confirmed by several tests, including quantitative NMR
- Product integrity – enhanced level of testing and characterization compared to standard grade
- Lot retention – ample supply of past lots retained to ensure future process testing
- Change management – Change Control Notification (CCN) service
- Consistent manufacture – batch-specific manufacturing documentation review
Specifica
Sulfo-NHS-LC-Biotin | |
Upon receipt store desiccated at -20° C. Product is shipped at ambient temperature. | |
Cell-Impermeant | |
Active Ester, Succinimidyl Ester, Sulfo-NHS Ester | |
Biotin | |
Biotin & Analogs |
1 g | |
Pierce™ | |
Amine | |
DMF (Dimethylformamide), DMSO (Dimethylsulfoxide), Water | |
Mid-length |
For Research Use Only. Not for use in diagnostic procedures.