missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Collibri™ Library Amplification Master Mix

Codice prodotto. p-8772008 Sfoglia Tutto Thermo Scientific Prodotti
Click to view available options
N. di reazioni:
50
250
Dimensione della confezione:
50 reazioni
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Codice prodotto. 15950523

Marca: Invitrogen™ A38539050

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Invitrogen™ Platinum SuperFi Library Amplification Master Mix is a ready-to-use solution designed for amplification of next-generation sequencing (NGS) libraries.

Master mix includes Platinum SuperFi™ DNA Polymerase in combination with a proprietary reaction buffer containing all necessary components for efficient library amplification with minimal bias and superior accuracy.

 Features include:

  • High fidelity—minimal error rates similar to PCR-free methods
  • High efficiency—high yields from minute input amounts
  • Minimal bias—uniform amplification regardless of GC content

 

  • Platinum SuperFi™ DNA Polymerase has both 5’ to 3’ polymerase and 3’ to 5’ exonuclease (proofreading) activities, but lacks 5’ to 3’ exonuclease activity. Exceptionally strong proofreading activity ensures amplification of NGS libraries with supreme sequence accuracy, producing NGS error rates similar to PCR-free methods
  • Proprietary reaction buffer contains all necessary PCR components and additives to enable efficient and uniform library amplification regardless of GC content, helping improve coverage across GC- and AT-rich sequences and other complex regions. High amplification efficiency enables the use of fewer cycles to achieve the desired yield
  • Platinum hot-start technology inhibits DNA polymerase activity at ambient temperatures, allowing room temperature reaction setup and storage of pre-assembled PCR reactions for up to 24 hours prior to the PCR
  • Platinum SuperFi™ Library Amplification Master Mix is supplemented with an inert blue dye that provides a visual aid when pipetting and decreases the risk of pipetting errors during reaction setup
  • For increased convenience, we offer Platinum SuperFi™ Library Amplification Master Mix with Primer Mix, where the master mix is supplied with a 10X Primer Mix that targets the P5 and P7 regions of Illumina adapters and that is supplemented with an inert yellow dye. Mixing both components in a PCR reaction turns the final PCR solution green, further decreasing the risk of pipetting errors
TRUSTED_SUSTAINABILITY

Specifica

Content And Storage 1 x 1.25 mL Collibri Library Amplification Master Mix (2X)

Store at -20°C.
Tipo di campione DNA, RNA
Fedeltà (rispetto a taq) ≥100
Prestazioni PCR arricchito GC High
Da utilizzare con (applicazione) Sequencing
Hot Start Built-In Hot Start
N. di reazioni 50
Sporgenza Blunt
Linea di prodotti Collibri
Product Type Library Amplification Master Mix
Quantità 50 reactions
Reaction Format SuperMix or Master Mix
Shipping Condition Dry Ice
Durata del test Standard
Forma Liquid
Vedi altri risultati Mostra meno risultati

For Research Use Only. Not for use in diagnostic procedures.

Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato