Learn More
Socorex™ Filtro di protezione per ugello in polietilene
Use the Socorex™ Polyethylene Nozzle Protection Filter to guard against liquid overflow or aerosol contamination. The filter is interchangeable, non-sterile and non-autoclavable.
Marca: Socorex™ 322.810
Dettagli aggiuntivi : Peso : 0.00630kg
Descrizione
- Non-autoclavable
- Non-sterilised
- 100 Pack
- Polyethylene (PE)
Specifica
No | |
Filtro protezione ugello | |
Acura™ manual 810 (for autoclavable Acura 810 model only) |
Polyethylene | |
Protezione ugello | |
Confezione da 100 |