Learn More
Masterflex™ Raccordo scanalato per connettore riduttore in polietilene
Codice prodotto. 11800220
Click to view available options
Da utilizzare con:
1/2 x 1/4 in. tubing I.D.
1/4 x 3/16 in. tubing I.D.
3/8 x 1/4 in. tubing I.D.
5/8 x 1/2 in. tubing I.D.
5/8 x 3/8 in. tubing I.D.
Dimensione della confezione:
pezzi 10
Codice prodotto. 11800220
Marca: Masterflex™ 3062229
Combine these simple, economical HDPE reducing connector with flexible tubing for use in your low-pressure applications (generally less than 30 psi). HDPE Reducing Connector Barbed Fittings slide into soft tubing. Tubing elasticity holds the tubing onto the fitting for low-pressure installations.
  • Designed for quick and easy assembly without tools
  • Very Good chemical resistance
  • Temp range: - 53 to 65°C
  • Sterilize by ethylene oxide
Specifica
Materiale Polyethylene
Da utilizzare con 1/4 x 3/16 in. tubing I.D.
Quantità confezione Confezione da 10
Product Type Adattatore portagomma