Learn More
Macherey-Nagel™ Polygoprep™ 1000 to 12
Marca: Macherey-Nagel™ 711035.1000
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
10 to 15 μm | |
POLYGOPREP 1000 to 12 | |
1 kg | |
Polygoprep |
1000 Å | |
2 to 8 | |
'POLYGOPREP | |
Liquid Chromatography |