Learn More
Brady™ Polypropylene Warning Sign
Marca: Brady™ 223439
Questo articolo non è restituibile.
Consulta la politica di reso
Specifica
ISO 7010 | |
Polypropylene | |
850 N/100 mm | |
100 mm | |
100 mm | |
Triangular | |
Warning Sign |
PIC 300-TRI 100-B7527 | |
1 Ea. | |
Signs can be used for a wide variety of applications both indoors and outdoors | |
0.369 mm | |
-40°C to 120°C | |
Warning of Flammable Materials | |
Black on Yellow |