Learn More
InterLab™ Supply PolySeed™ Microbiol BOD Capsules
PolySeed™ Microbiol BOD Capsules - Easy to use capsules contain a special blend of microbial cultures that provide a uniform standard for BOD analysis.
Marca: InterLab™ Supply 53200-33
Dettagli aggiuntivi : Peso : 0.06800kg
Descrizione
- Fresh consistent seeds each time
- One capsule performs approximately 100 tests
- Easy to use
Specifica
BOD analysis | |
One capsule performs 100 tests | |
50 capsules per bottle | |
BOD microbial BOD capsules |