Learn More
Greiner Bio-One Micropiastra per colture cellulari da 1536 pozzetti in polistirene con coperchio
Designed with a rounded square profile for flexible working volume combined with reduced wicking and bubbling. Greiner Bio-One™ Polystyrene 1536 Well HiBase Microplates with Lid are ideally suited for top reading instruments, fluorescence measurements and, due to hydrophobic nature of surface, non polar proteins and peptides.
Marca:  Greiner Bio-One 782086
Dettagli aggiuntivi : Peso : 0.80000kg
Codice prodotto. 11878990
  • 4645.00€ / pezzi 40
Spedizione stimata: 13-08-2024
per visualizzare lo stock disponibile
Aggiungi al carrello
Descrizione
Descrizione
  • Clear polystyrene construction for optimum visibility
  • Can be traced to production through defined LOT number system
  • Free of detectable DNase, RNase and human DNA
  • Non-pyrogenic, non-cytotoxic
  • Manufactured under DIN ISO 9001 guidelines
  • Alphanumeric well coding
  • Compatible with automated systems
  • Square wells with rounded corners combines advantages of square well (i.e. flexible working volume of 3μ to 10μ with the advantages of a round well (reduced wicking and bubbling) 
Specifica
Specifica
DIN ISO 9001
Standard
32 x 48
Polystyrene
3 to 10 μL
Tissue Culture
Flat Bottom (F-well)
Confezione da 40
Free of detectable DNase, RNase, human DNA, non-pyrogenic
With Lid
Nero
1536
Sterile
Da 3 a 10 μl