Learn More
LABPLAS Polyurethane Sani-Sponge with Dey-Engley Neutralizing Broth
Synthetic polyurethane sponge for sampling surfaces, with buffer to neutralize disinfectant.
Marca: LABPLAS KSS-67810-DE
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- Polyurethane Sani-Sponge
- Saturated with Dey-Engley neutralizing broth
- Dimensions: 160 x 229 mm
- Holds: 600 mL
- Option to include gloves with kit
Specifica
Hygiene Sponge Sampling Kit | |
Neutralizers | |
Sterile | |
None |