Learn More
Epredia™ PrintMate™ Embedding Cassettes
Sfoglia Tutto Epredia Prodotti
Click to view available options
Colore:
Aqua
Beige
Blue
Gray
Green
Lavender
Light Green
Lilac
Orange
Pink
Tan
White
Yellow
Dimensione della confezione:
pezzi 1000
pezzi 750
Quantità:
10 x 75 cassettes (750)/cs
1000
4 x 250 cassettes (1000)/cs
Chiusura inclusa:
No lids
Separate lids included
Descrizione
Designed for performance and user convenience, PrintMate Embedding Cassettes have been optimized for superior workflow and printing with the PrintMate Classic and PrintMate AS printing systems. The even, textured printing face provides the optimal surface for high-quality printing every time. For additional information, the right and left sides are textured for clear handwriting with suitable pens. The lid catches have been adjusted for ease of use with low opening and closing force, while still giving the security needed while processing.
Available in tubes for automatic printing or loose for manual use, and in twelve colors for enhanced workflow identification.
PrintMate Embedding Cassettes are also available without lids, for economical use with reuseable metal lids.
Available in tubes for automatic printing or loose for manual use, and in twelve colors for enhanced workflow identification.
PrintMate Embedding Cassettes are also available without lids, for economical use with reuseable metal lids.
Specifica
Specifica
| Stile pori | Round Pore |
| Compatibilità con PrintMate | Yes |
| Colore | Pink |
| Product Type | Embedding Biopsy Cassette |