missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pro-Lab Diagnostics™ Prolex™ Complete Streptococcal Grouping Latex Kit
Provides a rapid method for the serological identification of groups A, B, C, D, F and G of the Lancefield groups of streptococci grown on agar plates
Marca: Pro-Lab Diagnostics™ PL030
Includes:
Specific Latex Suspensions for Groups A-G (3mL each); Polyvalent Positive Control (2mL); Extraction Reagent 1 (3.2mL); Extraction Reagent 2 (3.2mL); Extraction Reagent 3 (2 x 8.0mL); Agglutination Cards (48); Mixing Sticks (350)
Requires :
- Inoculating loops
- Pasteur pipets
- Borosilicate glass test tubes (12mm x 75mm)
- Timer
Questo articolo non è restituibile. Consulta la politica di reso
Descrizione
- Offers a unique method for grouping Lancefield beta-haemolytic Streptococci
- Utilizes modified acid extraction reagents and blue carboxylated latex particles
- Results are obtained within one minute at room temperature
- All kit reagent labels and caps are color coded for ease of identification, and presented in easy-to-use, convenient dropper bottles
All components are also available individually
Specifica
2°C to 8°C | |
Complete Streptococcal Grouping Latex Kit | |
60 Tests |
99% | |
3mL Specific Latex suspensions for groups A, B, C, D, F and G, 2mL Polyvalent positive control, 3.2mL extraction R1, 3.2mL extraction R2, 2 x 8mL extraction R3, 48 Agglutination cards and 350 mixing sticks | |
1 |