Learn More
Pro-Lab Diagnostics™ Prolex™ Streptococcal Grouping Latex Kits
For grouping Lancefield betahaemolytic Streptococci
Marca: Pro-Lab Diagnostics™ PL032
This item is not returnable.
View return policy
Description
- Instant results obtained at room temperature
- Advanced extraction process using modified nitrous acid reagents
- Enhanced results with blue carboxylate-modified microparticles (CM-MP)
- As few as one colony required
- Colour coded reagents and vials for ease of selection
- Available individually
Specifications
2°C to 8°C | |
Group B Latex Reagent | |
3mL Blue Latex particles coated with purified Rabbit Antibodies | |
3 mL |