Learn More
Pro-Lab Diagnostics™ Prolex™ Streptococcal Grouping Latex Kits
For grouping Lancefield betahaemolytic Streptococci
Marca:  Pro-Lab Diagnostics™ PL033
Dettagli aggiuntivi : CAS : 26628-22-8 Peso : 0.80000kg Transport : UN number : 3316 Chem class : 9 Pack group : II
Codice prodotto. 11929044
  • 41.35€ / pezzi 60
Spedizione stimata: 14-10-2024
per visualizzare lo stock disponibile
Descrizione
Descrizione
  • Instant results obtained at room temperature
  • Advanced extraction process using modified nitrous acid reagents
  • Enhanced results with blue carboxylate-modified microparticles (CM-MP)
  • As few as one colony required
  • Colour coded reagents and vials for ease of selection
  • Available individually
Specifica
Specifica
2°C to 8°C
Group C Latex Reagent
3mL Blue Latex particles coated with purified Rabbit Antibodies
3 mL