Learn More
Hoefer™ High-Voltage Lead Set
Accessory for Electrophoresis Unit
Marca:  Hoefer™ SE6056-HV
Dettagli aggiuntivi : Peso : 0.40000kg
Codice prodotto. 15508991
  • 172.24€ / 1 set
Spedizione stimata: 08-10-2024
per visualizzare lo stock disponibile
Aggiungi al carrello
Descrizione
Descrizione
  • High-voltage Lead Set (1 red and 1 black)
  • Female end is for use with Hoefer safety lids only
  • Male end is sheathed and may require an adaptor to fit power source from another manufacturer
Specifica
Specifica
High Voltage Lead Set
One red, one black