Learn More
Thermo Scientific™ KingFisher™ Presto Purification System
Use the Thermo Scientific KingFisher Presto as part of an automated purification system with a liquid handler for plate filling in high-throughput laboratories.
Marca:  Thermo Scientific™ 24078840
Dettagli aggiuntivi : Peso : 1.00000kg
Codice prodotto. 15851647
  • 4145.00€ / Pezzo
Spedizione stimata: 12-08-2024
per visualizzare lo stock disponibile
Descrizione
Descrizione
Use the Thermo Scientific KingFisher Presto as part of an automated purification system with a liquid handler for plate filling in high-throughput laboratories. Utilizing proven KingFisher magnetic particle-based technology, the KingFisher Presto provides high-quality yields of target nucleic acids and proteins for biopharma, biotech and research.
Features:
• Purify 96 or 24 samples at a time
• Volume range of 50–5000 μL
• Minimized size for easy fitting to automation systems
• Heating capability for application requiring elevated temperatures
• Easy connection to several liquid handling instruments
• Flexible system for wide variety of applications
Specifica
Specifica
200 to 5000 μL
KingFisher Presto 24 Deep Well head
24