Learn More
Thermo Scientific Chemicals PTFE Tweezers-Forceps
Click to view available options
Lunghezza (sistema metrico):
100 mm
200 mm
Stile punta:
Sharp
Square
Dimensione della confezione:
Pezzo
Descrizione
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- PTFE
Specifica
Specifica
| Product Type | Tweezers-Forceps |
| Materiale | PTFE |
| Stile punta | Square |
| Lunghezza (sistema imperiale) | 7.8 in. |
| Lunghezza (sistema metrico) | 200 mm |