Learn More
Thermo Scientific™ Purified Water for Microbiology
Perform sample dilution for microbial examination or ensure sterility of your devices and equipment with Purified Water for Microbiology.
Marca: Thermo Scientific™ BO0209C
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
Perform sample dilution for microbial examination or ensure sterility of your devices and equipment with Thermo Scientific™ Oxoid™ Purified Water. The bottles come with screw cap closure for ease of operation.
Use Purified Water as a diluting and rinsing fluid.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifica
Colorless, clear | |
Purified Water for Microbiology | |
United Kingdom | |
5.0g + 0.3g | |
24 x 5 mL in BIJOU |
2 to 25°C | |
Bijou bottles | |
Purified Water | |
Twenty-four bijou bottles with screw cap closures in a box. |