Learn More
Thermo Scientific Chemicals Quartz, cover slip for microscope slide, fused, 25.4x25.4x0.15-0.25mm (1.0x1.0x0.006-0.01in)
Codice prodotto. p-6685053
Click to view available options
Quantità:
1/Ea.
5/Ea.
Dimensioni (L x P):
25.4 mm
25.4 x 25.4 mm
Dimensione della confezione:
Pezzo
pezzi 5
Codice prodotto. 15473885
Marca: Thermo Scientific Chemicals 043210.KG
Solo null disponibili
Aggiungi al carrello
For use with microscope quartz slides made by fusing naturally occurring crystalline silica
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Cover slips for use with microscope quartz slides
TRUSTED_SUSTAINABILITY
Specifica
Spessore (sistema metrico) 0.15 to 0.25 mm
Descrizione Cover Slips for use with Microscope Quartz Slide
Quantità 1/Ea.
Dimensioni (L x P) 25.4 x 25.4 mm