Learn More
Thermo Scientific Chemicals Quartz disc, fused, 50.8mm (2.0in) dia x 3.18mm (0.13in) thick
Click to view available options
Diametro (sistema metrico):
50.8 mm
76.2 mm
Spessore:
1.59 mm
3.18 mm
Dimensione della confezione:
Pezzo
Descrizione
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifica
Specifica
| Product Type | Mineral, Quartz Disc |
| Descrizione | Ground and polished |
| Diametro (sistema metrico) | 50.8 mm |
| Spessore | 3.18 mm |