Learn More
Colonne LC Quickfit™
Click to view available options
Lunghezza (sistema metrico):
200 mm
300 mm
400 mm
500 mm
Diametro (sistema metrico) interno:
10 mm
20 mm
26 mm
40 mm
Tipi di connettore:
14/23 Socket
19/26 Socket
24/29 Socket
40/38 Socket
Include:
GP Rotaflo 3mm Bore Stopcock
GP Rotaflo 6mm Bore Stopcock
Dimensione della confezione:
Pezzo
Descrizione
Manufactured from Pyrex™ borosilicate glass. Quickfit™ Chromatography Column with Rotaflo™ Stopcock includes a 0 porosity chemically resistant glass sintered disc. PTFE Stopcock provides fine control of eluent flow. Ground glass socket accepts Quickfit™ reservoirs.
- With PTFE Rotaflo™ Stopcock for fine control of eluent flow
- Chemically resistant glass sintered disc - porosity 0
- Ground glass socket accepts Quickfit™ reservoirs
- Manufactured from Pyrex™ borosilicate glass
Specifica
Specifica
| Quantità | 1 Pack |
| Diametro (sistema metrico) interno | 10 mm |
| Tipo | Chromatography Column |
| Lunghezza (sistema metrico) | 200 mm |
| Materiale | Pyrex™ Borosilicate Glass |
| Tipi di connettore | 14/23 Socket |
| Include | GP Rotaflo 3mm Bore Stopcock |