Learn More
Quickfit™ Dilatometer Body
For measuring the expansion in volume of edible oils and fats over a temperature range within which melting takes place
Marca: Quickfit WE12/1
850.25 EUR valido fino al 2024-12-31
Usa il codice promo "23259" per avere il tuo prezzo promozionale.
Dettagli aggiuntivi : Peso : 0.07000kg
Descrizione
- Dilatometer body is graduated with an arbitrary scale, from 0 to 0.9mL
- Graduated at every 0.005mL with clear figures every 0.05mL
- Chamber is filled with oil or fat and the meniscus should be settled on zero when the stopper is in place at starting temperature
Specifica
Dilatometer | |
For measuring the expansion of edible oils and fats over the temperature range within which melting takes place |