missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human ADRA1D (NP_000669) VersaClone cDNA
Alpha-1D Adrenergic Receptor
Marca: R&D Systems™ RDC0255
Dettagli aggiuntivi : Peso : 0.00970kg
Descrizione
The Human ADRA1D (NP_000669) VersaClone cDNA clone contains the complete ORF for alpha-1D Adrenergic R/ADRA1D, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The alpha-1D Adrenergic R/ADRA1D insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.Order Info
Ships at ambient temperature.
Specifica
hADRA1D | |
10 μg at 0.2 μg/μL | |
1732bp | |
Human |
NP_000669 | |
cDNA is provided in 10mM Tris-Cl, pH 8.5 | |
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
For Reasearch Use Only; Not for Use in Humans