Learn More
R&D Systems™ Human GPR135 (NP_072093) VersaClone cDNA
G Protein-coupled Receptor 135
Marca: R&D Systems™ RDC0831
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
The Human GPR135 (NP_072093) VersaClone cDNA clone contains the complete ORF for GPR135, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The GPR135 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.Order Info
Ships at ambient temperature.
Specifica
hGPR135 | |
10 μg at 0.2 μg/μL | |
1498bp | |
Human |
NP_072093 | |
cDNA is provided in 10mM Tris-Cl, pH 8.5 | |
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
For Reasearch Use Only; Not for Use in Humans