Learn More
R&D Systems™ MagCellect Mouse Hematopoietic Cell Lineage Depletion Kit
Designed to isolate hematopoietic non-lineage committed cells (lineage-negative) from mouse bone marrow
Marca:  R&D Systems™ MAGM209
Codice prodotto. 18471360
  • 777.00€ / 1 set
Esplora altre offerte speciali
Questo articolo non è restituibile. Consulta la politica di reso
Descrizione
Descrizione
Resulting cell population is highly enriched for CD117+ cells (from 40-70% depending on mouse strain) with less than 5% residual lineage positive cells
Isolated HSCs are untouched by beads or antibodies
Negative selection reduces sources of experimental variation
Lineage committed cells (lineage positive) targeted for magnetic depletion include T cells, B cells, NK cells, monocytes/macrophages, granulocytes, and erythrocytes
Specifica
Specifica
Store the unopened product at 2°C to 8°C. Do not use past expiration date.