Learn More
R&D Systems™ Mouse LPAR1/LPA1/EDG-2 (NP_034466) VersaClone cDNA
Lysophosphatidic Acid Receptor 1
Marca:  R&D Systems™ RDC0565
Codice prodotto. 18441920
  • 387.00€ / 10µg
Esplora altre offerte speciali
Questo articolo non è restituibile. Consulta la politica di reso
Descrizione
Descrizione
The Mouse LPAR1/LPA1/EDG-2 (NP_034466) VersaClone cDNA clone contains the complete ORF for LPAR1/LPA1/EDG-2, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The LPAR1/LPA1/EDG-2 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Order Info
Ships at ambient temperature.
Specifica
Specifica
mLPAR1
10 μg at 0.2 μg/μL
1108bp
Mouse
NP_034466
cDNA is provided in 10mM Tris-Cl, pH 8.5
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
10 μg
For Reasearch Use Only; Not for Use in Humans