Learn More
R&D Systems™ Proteome Profiler Human Chemokine Array Kit
Click to view available options
Quantità:
1 Kit
Dimensione della confezione:
1 set
Descrizione
The Proteome Profiler Human Chemokine Array Kit contains 4 membranes - each spotted in duplicate with 31 different chemokine antibodies.
Specifica
Specifica
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Quantità | 1 Kit |