missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Donkey IgG (H+L) Secondary Antibody, FITC, Invitrogen™
Rabbit Polyclonal Secondary Antibody
Marca: Invitrogen PA128588
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Anti-Donkey secondary antibodies are affinity-purified antibodies with well-characterized specificity for equine (Donkey) immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.Specifica
Donkey IgG (H+L) | |
Polyclonal | |
FITC | |
Rabbit | |
IgG fraction | |
RUO | |
Donkey | |
Liquid |
Flow Cytometry, Immunocytochemistry | |
2 mg/mL | |
PBS with no preservative; pH 7.2 | |
donkey whole molecule | |
1 mg | |
Secondary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto